Constant Climate Chambers, Memmert | Avantor
Constant Climate Chambers, Memmert
Supplier:   Memmert
Constant Climate Chambers, Memmert
Supplier:  Memmert
Certificates
Constant Climate Chambers, Memmert
Constant Climate Chambers, Memmert
1 Options Available Below
Product Details & Documents
The HPP eco constant climate chambers are energy efficient, precise and eco-friendly.
  • For temperatures with humidity from 5 up to 70 °C
  • Humidity range 10 to 90% RH
  • Energy-saving Peltier heating/cooling system integrated in the rear
  • Programmable with internal data logging via included AtmoCONTROL Software
These climate chambers are programmable models with Peltier-technology that seamlessly heats and cools, these are are 115 V models with TwinDISPLAY digital controller.
Certifications: Each individual appliance is tested according to the requirements of DIN 12 880: 2007-05 and units are CE marked.
Delivery information: Supplied with 2 stainless steel grid shelves, 2.5 L water tank, operating manual, AtmoCONTROL Software with user manual, calibration certificate, power cord. Other options and accessories are not included and may affect lead time.
Packaging: Ships on pallet.
Product Family Options
Informations sur le produit
  • Désignation
    Modèle
    Volume
    No. of Shelves
    Interior Dimensions
    Exterior Dimensions
    Plage de température
    Electrical
    Availability
    Price
  • HPP110eco climatic test chamber
    HPP110ECO
    108 L
    2 (5)
    560×400×480 mm (22×15.7×18.9")
    745×555×864 mm (29×22×34")
    5…70 °C
    115 V
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26,540.60
    $26,540.60
    Each
       $26,540.60
    Supplier Number: HPP110ECO
  • HPP260eco climatic test chamber
    HPP260ECO
    256 L
    2 (9)
    640×500×800 mm (25×19.7×31.5")
    824×655×1183 mm (32.4×25.7×46.6")
    5…70 °C
    115 V
    Discontinued
    Supplier Number: HPP260ECO P1
  • HPP410eco climatic test chamber
    HPP410ECO
    384 L
    2 (14)
    640×500×1200 mm (25.2×19.7×47.2")
    824×655×1720 mm (32.4×25.7×67.7")
    5…70 °C
    115 V
    Discontinued
    Supplier Number: HPP410ECO P1
  • Climatic test chamber
    HPP750ECO
    749 L
    2 (14)
    1040×600×1200 mm (40.9×23.6×47.2")
    1224×755×1720 mm (48×29.7×67.7")
    5…70 °C
    115 V
    Discontinued
    Supplier Number: HPP750ECO P1