Centrifuge Tubes, RNase, DNase and Pyrogen Free, Globe Scientific
76478-224
- Cap Material:HDPE
- Cap Type:Screw cap
- Sterility:Sterile
- Material:PP
- Cat. no.:76478-224
- Packaging:25/bag, 20 bags/case
- Cap Color:Blue
These tubes are produced from polypropylene.
- HDPE cap
- Gamma sterile, RNase/DNase free
- Max spin rate: 1000×g
They feature printed graduations and a white writing space.