BCYE Agar with GVPC, without Cysteine
Catalog # CA76359-398
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Plate
- Description:Prepared media (BCYE agar with GVPC, without cysteine)
- Size:10/pack
- Cat. no.:CA76359-398
- Tube Size:15×100 mm
Specifications
About this item
Buffered charcoal yeast extract (BCYE) agar formulations are recommended for use in the cultivation and primary isolation of Legionella spp. in water and other samples suspected of harboring the bacteria.
- Optically clear polystyrene for distortion free viewing
- Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
- Packaged in sleeves of 10 plates in a "breathable" bag that prevents buildup of condensation and excess moisture