Sterilization Wraps, Heavy Duty for Steam EtO or Gamma Irradiation, Blue, Keystone Cleanroom Products
Fournisseur: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificats
À propos de cet article
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an effective microbial barrier from external sources of contamination.
- Treated to impart water and alcohol repellency
- Sufficient microbial barrier; superior bacterial barrier properties compared to linen
- Sterilisation tapes adhere firmly and remove easily
- Ideal choice for for light trays, small basins and towels
- Resists tearing and punctures
- Does not contain any natural latex rubber
- Processed and packaged in ISO certified cleanrooms
These versatile wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non-woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
These wraps are wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder and are designed for steam sterilization; but also compatible with EtO or gamma irradiation.
Certifications: Certificates of Conformance with each shipment.
Frequently Bought Together