J.T.Baker® Plate 384
   Sustainable Options Available
J.T.Baker® Plate 384
   Sustainable Options Available
Certificates
J.T.Baker® Plate 384
2 Options Available Below
Product Details & Documents
Available in PS and PP for automated screening and storage systems.
  • Alphanumeric code to assist sample identification
  • Square wells for increased volume
  • V-bottom for high sample recovery
  • High frame (3 mm) for stable and slip-free stacking
  • Higher well rims avoid cross-contamination and allow better closure with adhesive films
J.T.Baker® Plate 384 is ideal for small sample volumes.
Specifications for Product Family
Specifications
  • Catalog No:
  • 43004-0120
  • 43004-0200
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Specifications
Product Family Options
Product Information
  • Désignation
    Well Volume
    Working Volume
    Color
    Sterile
    Availability
    Price
  • PP plates
    120 µl
    5 - 110 µl
    Transparent
    -
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $1,014.15
    $1,014.15
    Case of 60
       $1,014.15
    Supplier Number: 43004-01201
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43004-0120
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • PP plates
    200 µl
    5 - 180 µl
    Transparent
    -
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $687.96
    $687.96
    Case of 40
       $687.96
    Supplier Number: 43004-02001
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43004-0200
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact