J.T.Baker® Robotic Tips Type MP
Catalog # 49209-0105
Supplier: Ritter, part of Avantor
Specifications
- Volume:1000 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Tip Color:Black
- Packed:8× 5 Racks, 96 each, blistered
- Cat. no.:49209-0105
- Sterile:+
Specifications
About this item
Suitable for use with Roche MagNA Pure instruments. Available in conductive version, with filter.
- Manufactured under cleanroom conditions (cleanroom Class 8)
- State of the art quality control
- Barcoded trays for automated identification
- Free of DNA, DNase, RNase and PCR inhibitors
- Ideal for high throughput analysis
- Packed in blisters
The validation of the tips on the instrument for specific applications is within the responsibility of the user.