CompactDry™ S. aureus (X-SA), Hardy Diagnostics
Catalog # CA10145-970
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:CompactDry™ S. aureus (X-SA) Cassettes
- Size:240 plates/pack
- Storage temperature:2 to 30 °C away from direct light
- Cat. no.:CA10145-970
- Supplier No.:XSA100
Specifications
About this item
CompactDry™ X-SA is a ready-to-use test method recommended for the isolation and enumeration of Staphylococcus aureus in raw materials, finished products, or on environmental surfaces pertaining to food and related industries.
- Sturdy frame, stackable packaging
- Self-diffusing media
- Ability to subculture without damage to colonies
- No leakage
- Easy to label
CompactDry™ X-SA for the detection and enumeration of Staphylococcus aureus.
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: This product is not intended to be used for the diagnosis of human disease. Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.