ContactUsLinkComponent
FLM300 Fluorophos ALP Test System, Advanced Instruments
FLM300 Fluorophos ALP Test System, Advanced Instruments
Catalog # 76474-156
CAS Number:  
undefined
FLM300 Fluorophos ALP Test System, Advanced Instruments
Catalog # 76474-156
Supplier Number:  FLM300
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    FLM300 fluorophos ALP test system
  • Cat. no.:
    76474-156
Specifications
About this item
The Fluorophos ALP assay is a rapid test for verifying proper pasteurization of dairy products.
  • Fast and accurate results
  • Globally proven
  • Reduced production liability
Alkaline Phosphatase (ALP), found naturally in raw milk, is destroyed at temperatures slightly above the temperature necessary to destroy microorganisms pathogenic to humans. Measurement of residual phosphatase after pasteurization offers a means of verifying the products are heated to the proper temperatures and are not contaminated with raw milk. The Fluorophos ALP assay measures the ALP activity in dairy products, allowing the user to monitor the completeness of pasteurization and detect raw milk contamination.