Single-Sample Cryoscope, Model 4250, Advanced Instruments | Avantor
Single-Sample Cryoscope, Model 4250, Advanced Instruments
Single-sample cryoscope, Model 4250
Catalog # 76474-142
Fournisseur #:  4250
Single-Sample Cryoscope, Model 4250, Advanced Instruments
Catalog # 76474-142
Single-Sample Cryoscope, Model 4250, Advanced Instruments
Single-Sample Cryoscope, Model 4250, Advanced Instruments
Single-sample cryoscope, Model 4250
Price
$34,271.89
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The single-sample cryoscope, model 4250 is a fast, reliable, and precise system for milk analysis.
  • Low maintenance
  • On-board printer for better traceability
  • Rapid and stable automated calibration
The model 4250 employs the industry preferred freezing point depression (FPD) method for determination of added water to milk. It allows your plant to manage a premium milk supply, ensure quality products and improve your operating efficiency.
Spécifications
  • Désignation:
    Single-sample cryoscope, Model 4250
  • Cat. No.:
    76474-142
  • Sample volume:
    2.0 or 2.5 ml
  • Linearity:
    Less than ±0,5% from straight line (96-wel plate)