KoolPlate™ Cooling Tray, Simport | Avantor
KoolPlate™ Cooling Tray, Simport
KoolPlate™ 6 cooling trays with 1 insulator base, 11×5.5×1"
Catalog # 76479-186
Supplier:   Simport Scientific
Fournisseur #:  M479-24
KoolPlate™ Cooling Tray, Simport
Catalog # 76479-186
Supplier:  Simport Scientific
KoolPlate™ Cooling Tray, Simport
KoolPlate™ Cooling Tray, Simport
KoolPlate™ 6 cooling trays with 1 insulator base, 11×5.5×1"
Price
$371.06
Log in pour la disponibilité
Case of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Simport KoolPlate™ is ideal for chilling and preparing paraffin blocks, for sectioning at the microtome or for performing other laboratory procedures.
  • For chilling and preparing paraffin blocks
  • Keeps paraffin blocks cold up to 4 hours
  • Tray made of high impact polystyrene for longevity
  • Insulator base made of styrofoam
The KoolPlate™ freezes overnight (16 hours) in a conventional freezer and the surface remains cold for up to 4 hours when used with the supplied styrofoam insulator base placed under the KoolPlate™. It will prolong cooling and prevent moisture accumulation. It is made of thick wall high impact polystyrene in order to ensure a sturdy construction that will last for years.
The KoolPlate™ is stackable, a great space-saving feature. Warning–Not for use with organic solvents such as toluene, xylene, acetone, etc.
 : Not for use with organic solvents such as toluene, xylene, acetone, etc.
Spécifications
  • Désignation:
    KoolPlate™ 6 cooling trays with 1 insulator base
  • Dimensions:
    11×5.5×1"
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Styrofoam insulator base only, 6/case
    Styrofoam insulator base only, 6/case
    76479-188
    Log in pour la disponibilité
    Case of 6 - $71.01