TE Buffers 100X, G-Biosciences
Catalog # CA82023-084
Supplier: G-Biosciences
CAS Number:
Specifications
- Description:TE Buffer, 100X
- Size:100 ml
- Cat. no.:CA82023-084
- pH:8.0
- Supplier No.:R003
Specifications
About this item
TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.