ContactUsLinkComponent
VICTOR® Nivo™ Plate Readers, Multi-Mode
VICTOR® Nivo™ Plate Readers, Multi-Mode
Certificates
undefined
VICTOR® Nivo™ Plate Readers, Multi-Mode
Specifications
  • Catalog No:
  • CA76458-362
  • CA76458-360
  • CA76458-358
  • Model:
  • Victor Nivo 6T
  • Victor Nivo 5T
  • Victor Nivo 3T
  • Measuring modes:
  • Absorbance, Luminescence, Fluorescence, TRF/TR-FRET, FP, Alpha
  • Absorbance, Luminescence, Fluorescence, TRF/TR-FRET, FP
  • Absorbance, Luminescence, Fluorescence
Specifications
Related Information
Frequently Bought Together