ContactUsLinkComponent
VWR® PBS-T (Phosphate Buffered Saline - Tween-20) Buffer Powder Packs
Catalog # 76371-736
Supplier:  VWR International
CAS Number:  
undefined
VWR® PBS-T (Phosphate Buffered Saline - Tween-20) Buffer Powder Packs
Catalog # 76371-736
Supplier:  VWR International
CAS Number:  
Specifications
  • Description:
    VWR® PBS-T Buffer Powder
  • Size:
    100 pks
  • Cat. no.:
    76371-736
  • pH:
    7.2 - 7.6
  • Formulation:
    137 mM NaCl, 2.7 mM KCl, 10mM phosphate, 0.05% Tween-20
Specifications
About this item
VWR® Buffer Powder Packs can save time and money, as there is no need to measure and weight many separate chemical components which is required when making buffers from scratch.
  • Buffer powder quickly dissolves to make 1L of 1X solution
  • No weighing or measuring of separate components
  • No pH adjustments required
  • Economical shipping, long shelf life
  • Quality and consistency guaranteed
VWR® Pre-measured buffer packs are extremely easy to use and are formulated so each pack makes 1 Liter of 1X buffer. Simply tear open a pack and mix completely with 1 liter of distilled water.
Shipping costs are also minimal, as each case of 50 or 100 packs is compact and weighs only 2.5 Lbs.
Certifications:
COA available for each lot number.
Packaging: Each case of buffer packs is packaged in a convenient dispenser box, which takes up very little lab space. Included in each case is an instruction insert, and a roll of stickers to label each container of prepared buffer solution.