Unisette™ Tissue Processing and Biopsy Cassettes, Simport
À propos de cet article
Our extremely popular Unisette™ histology cassette is now offered with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
- Made of acetal
- 45° anterior writing area
- One-piece integral lid that can be opened and closed up to three times without breakage
- Secure locking system without danger of specimen loss
- The biopsy cassette has 1 mm square openings to maximize fluid exchange
- Offered taped for use with Leica™ IPC printers
Molded from acetal, these cassettes keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap latch and hinge-lock design of the UNISETTE™ prevent early separation of base and lid and allows for one-hand operation.
22 Options Available Below
Product Details & Documents
Our extremely popular Unisette™ histology cassette is now offered with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
- Made of acetal
- 45° anterior writing area
- One-piece integral lid that can be opened and closed up to three times without breakage
- Secure locking system without danger of specimen loss
- The biopsy cassette has 1 mm square openings to maximize fluid exchange
- Offered taped for use with Leica™ IPC printers
Molded from acetal, these cassettes keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap latch and hinge-lock design of the UNISETTE™ prevent early separation of base and lid and allows for one-hand operation.
Product Family Options
Tissue cassettes in QuickLoad™ Stack (taped)
Informations sur le produit
- ColorUnitsAvailabilityPrice
- Supplier Number: M525-12T
- Supplier Number: M525-6T
- Supplier Number: M525-9T
- Supplier Number: M525-4T
- Supplier Number: M525-10T
- Supplier Number: M525-11T
- Supplier Number: M525-7T
- Supplier Number: M525-3T
- Supplier Number: M525-8T
- Supplier Number: M525-2T
- Supplier Number: M525-5T
Biopsy cassettes in QuickLoad™ Stack (taped)
Informations sur le produit
- ColorUnitsAvailabilityPrice
- Supplier Number: M526-12T
- Supplier Number: M526-6T
- Supplier Number: M526-9T
- Supplier Number: M526-4T
- Supplier Number: M526-10T
- Supplier Number: M526-11T
- Supplier Number: M526-7T
- Supplier Number: M526-3T
- Supplier Number: M526-8T
- Supplier Number: M526-2T
- Supplier Number: M526-5T