About this item
These cassettes keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage.
- Made of acetal
- 35° anterior writing area
- Large labeling area for easy identification
- One-piece integral lid that can be opened and closed up to three times without breakage
- Secure locking system without danger of specimen loss
- Compatible with Primera© Cassette printers
Molded from acetal, the one-piece integral lid eliminates the need for separate steel lids.The snap latch and hinge-lock design of the UNISETTE™ prevent early separation of base and lid and allows for one-hand operation. Each case contains three dispenser boxes of 500 cassettes.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.