Receipt Printer Paper, for Thermo Printer, for Wizard™ CompactDry&trade Reader
Replacement paper for Wizard™ CompactDry™ thermal printer, 3¹/₈"W × 230'L, 0.5" core
Catalog # CA76177-158
Supplier: HARDY DIAGNOSTICS CA
Supplier #: 120104
Receipt Printer Paper, for Thermo Printer, for Wizard™ CompactDry&trade Reader
Catalog # CA76177-158
Supplier: HARDY DIAGNOSTICS CA
Jump to:
Product Details & Documents
Replacement paper for CompactDry Wizard Thermal printer paper.
- Top quality paper deliver crisp, clear images
- Each paper rolls has an end of warning signal
Paper weight: 48 g
Industry: Food safety
Specifications
- Désignation:Replacement paper for Wizard™ CompactDry™ thermal printer
- Dimensions:3¹/₈"W × 230'L, 0.5" core