Eisco LabGlass® Boiling Flasks, Round Bottom with Threaded Joint
Supplier:   Wards
Eisco LabGlass® Boiling Flasks, Round Bottom with Threaded Joint
Supplier:  Wards
Certificates
Eisco LabGlass® Boiling Flasks, Round Bottom with Threaded Joint
Eisco LabGlass® Boiling Flasks, Round Bottom with Threaded Joint
10 Options Available Below
Product Details & Documents
High quality round bottom boiling flasks for chemistry classrooms and laboratories.
  • Made of borosilicate glass 3.3
  • Ground threaded joint for secure fit with other glassware
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Product Family Options
Informations sur le produit
  • Capacité
    Joint Size
    Availability
    Price
  • 50 ml
    14/23
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26.95
    $26.95
    Each
       $26.95
    Supplier Number: CH01002A
  • 50 ml
    19/26
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29.00
    $29.00
    Each
       $29.00
    Supplier Number: CH01002B
  • 100 ml
    14/23
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29.00
    $29.00
    Each
       $29.00
    Supplier Number: CH01002C
  • 100 ml
    19/26
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $30.05
    $30.05
    Each
       $30.05
    Supplier Number: CH01002D
  • 100 ml
    24/29
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $28.90
    $28.90
    Each
       $28.90
    Supplier Number: CH01002E
  • 250 ml
    19/26
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.50
    $31.50
    Each
       $31.50
    Supplier Number: CH01002F
  • 250 ml
    24/29
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.50
    $31.50
    Each
       $31.50
    Supplier Number: CH01002G
  • 500 ml
    19/26
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $44.40
    $44.40
    Each
       $44.40
    Supplier Number: CH01002H
  • 500 ml
    24/29
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $44.40
    $44.40
    Each
       $44.40
    Supplier Number: CH01002I
  • 1000 ml
    24/29
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.90
    $63.90
    Each
       $63.90
    Supplier Number: CH01002J