Corning® HYPERStack® Nest, Corning
HYPERStack® nest, Stainless steel
Catalog # 76478-956
Supplier: Corning
: 10047
Jump to:
Product Details & Documents
The HYPERStack® Nest accessory is designed for use with the Corning® HYPERStack® 36-layer cell culture vessel.
- Lightweight design: 2 lbs. (950 g)
- Simplify process consistency by consistently placing the vessel in the correct position to facilitate optimal fill and empty processes
- Designed to reduce the chance of pressurizing vessels and wetting filters
- Pivot angle precisely engineered in the Nest reduces lifting by the user and freeing hands to operate tubing clamps
- Made of stainless steel, the Nest can be sterilized and autoclaved
- Small footprint fits within the current HYPERStack® footprint and multiple Nests can be stacked for storage
The HYPERStack® Nest accessory is designed for use with the Corning® HYPERStack® 36-layer cell culture vessel to improve process control by consistently placing the vessel in the optimal positions and angles during use. The HYPERStack® Nest is designed to improve ergonomics by reducing multiple manipulations. Stainless-steel construction allows it to be sterilized and autoclaved, and the small and stackable footprint allows for convenient storage.
Spécifications
- Désignation:HYPERStack® nest
- Matière:Stainless steel