Leaf and Flower Press
Supplier: Ward's Science
Certificates
About this item
Extra strength for thicker samples.
- Small size for field collection
- Strapping provides heavier compression
Ideal for field work because of its portable size, 6×9". This plant press is frequently used by Ecologists to gather base-line data for impact studies. Also used by Botanists and Entomologists in host plant studies. Ruggedly constructed of ¼" plywood strengthened by hardwood side strips which provide uniform pressure.
Delivery: Plant press comes complete with 12 drying sheets, 12 corrugated ventilators, and two straps with buckles.
Frequently Bought Together