Corning® Closed System Cell Cubes, Corning
Corning® Closed System Cell Cubes, Corning
   Options durables disponibles
Supplier:   Corning
Corning® Closed System Cell Cubes, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
Corning® Closed System Cell Cubes, Corning
3 Options Available Below
Product Details & Documents
Our new offering of CellCube 10, 25, 100 modules, circulation loops, and connectors allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik® and MPC connectors.
  • Standardized closed system designs allow flexible setup of any CellCube module and aseptic connection to an oxygenator (sold separately)
  • Sterile and ready-to-use offerings eliminate the time and complexity of assembling in-house
  • Industry standard AseptiQuik® S/G connectors allow for robust, single-use aseptic connections
The Corning CellCube system provides a fast, simple, and compact, perfusion-based method for the mass culture of attachment-dependent cells. CellCube modules consist of a series of 10, 25, or 100 parallel, polystyrene plates, which translates to 8500, 21250 or 85000 cm² respectively, joined to create thin, sealed laminar flow spaces between adjacent plate.
When connected with an oxygenator and controller (sold separately), the system continually perfuses the cells with fresh medium for increased cell productivity. The CellCube system provides an environment that more closely simulates in vivo conditions than a static culture with its perfusion-based design. For additional information, refer to Corning® CellCube® System(CLS-BP-039).
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76474-044
  • 76474-046
  • 76474-048
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Cell Growth Area
    Availability
    Price
  • CellCube-10 layers, w/AC, 1/2
    8500 cm²
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $2,440.00
    $2,440.00
    Case of 2
       $2,440.00
    Supplier Number: 3231
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    76474-044
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • CellCube-25 layers, w/AC, 1/1
    21250 cm²
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $1,510.00
    $1,510.00
    Case of 1
       $1,510.00
    Supplier Number: 3232
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    76474-046
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • CellCube-100 layers, w/AC, 1/1
    85000 cm²
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $4,460.00
    $4,460.00
    Case of 1
       $4,460.00
    Supplier Number: 3233
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    76474-048
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • CellCube T-tubing, filter, 2-AC
    CellCube T-tubing, filter, 2-AC
    76474-050
    Log in pour la disponibilité
    Case of 4 - $2,160.00