Eisco LabGlass® Pear Shape Boiling Flasks, Interchangable Joint
Eisco LabGlass® Pear Shape Boiling Flasks, Interchangable Joint
Supplier:   Wards
Eisco LabGlass® Pear Shape Boiling Flasks, Interchangable Joint
Supplier:  Wards
Certificates
Eisco LabGlass® Pear Shape Boiling Flasks, Interchangable Joint
Eisco LabGlass® Pear Shape Boiling Flasks, Interchangable Joint
4 Options Available Below
Product Details & Documents
High quality pear shapeboiling flasks with ground, interchangable joint.
  • Made of high quality borosilicate glass 3.3
  • Interchangable, ground joint
  • Stopper not included
  • Pear shape design
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Product Family Options
Product Information
  • Capacité
    Joint Size
    Availability
    Price
  • 25 ml
    14/23
    Log in for availability
    Log in for availability
    Each
    Each - $9.55
    $9.55
    Each
       $9.55
    Supplier Number: CH0406A
  • 50 ml
    14/23
    Log in for availability
    Log in for availability
    Each
    Each - $10.05
    $10.05
    Each
       $10.05
    Supplier Number: CH0406B
  • 100 ml
    14/23
    Log in for availability
    Log in for availability
    Each
    Each - $12.00
    $12.00
    Each
       $12.00
    Supplier Number: CH0406C
  • 250 ml
    14/23
    Log in for availability
    Log in for availability
    Each
    Each - $14.15
    $14.15
    Each
       $14.15
    Supplier Number: CH0406D