Drop-Boy Dropper Bottle
Drop-Boy dropper bottle, 50 ml
Catalog # 470332-512
Supplier: Dynalon
Fournisseur #: 805224-0050
Jump to:
Product Details & Documents
Precise drop dosing.
- Bottle should have dropper tip cut off before first use
- Reclosable using screw-on cap
Drop-Boy is a HDPE dropping bottle with a LDPE drop dispenser and separate polypropylene screw cap.
Spécifications
- Capacité:50 ml
- Désignation:Drop-Boy dropper bottle