Mini Plant Press
À propos de cet article
For hobbyist or field collection.
- Sized for easy portability
- Elastic bands provide even compression
The solution to the field ecologist's or naturalist's problem of bringing back specimens for identification from the field. These very portable yet extremely functional mini-presses should be in every field biologists kit. As a teaching tool and for student use the mini-press is ideal.
: Plant press has two masonite boards, 12 cards and driers and two elastic bands.
9 Options Available Below
Product Details & Documents
For hobbyist or field collection.
- Sized for easy portability
- Elastic bands provide even compression
The solution to the field ecologist's or naturalist's problem of bringing back specimens for identification from the field. These very portable yet extremely functional mini-presses should be in every field biologists kit. As a teaching tool and for student use the mini-press is ideal.
: Plant press has two masonite boards, 12 cards and driers and two elastic bands.
Product Family Options
Press kit
Informations sur le produit
- DésignationLongueurWidthAvailabilityPrice
- Supplier Number: 506.3998.1
- Supplier Number: 506.3999.1
- Supplier Number: 506.3998.10
- Supplier Number: 506.3999.10
Refills
Informations sur le produit
- DésignationLongueurWidthAvailabilityPrice
- Supplier Number: 506.4024.100
- Supplier Number: 506.4026.100
- Supplier Number: 506.4021.100
- Supplier Number: 506.4025.100
- Supplier Number: 506.4027.100