MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
Highly alloyed SINGLE
Catalog # 76468-748
Supplier #:  80074-BAS-SS
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
Catalog # 76468-748
CAS Number:  
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
Highly alloyed SINGLE
CAS Number:  
Price
$926.15
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 µ diamond finish and carbon coated. Supplied in 2, 3, and 5 mm diameter and a location map for standard identification. They are supplied with fully authenticated certificates of analysis.
Spécifications
Spécifications
  • Désignation:
    Highly alloyed SINGLE