Rapid Response™ Fentanyl Test Strip
About this item
The Rapid Response™ Fentanyl (FYL) Test Strip is a rapid visual immunoassay for the qualitative detection of fentanyl at a cut-off of 200 ng/ml for liquid/powder or 20 ng/ml for urine.
- Parameter: FYL
- Calibrator: Fentanyl
- Cut-off: 200 ng/ml (Liquid/Powder), 20 ng/ml (Urine)
Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no control line).
Delivery information: Includes test strips, product insert.
Caution: This test does not evaluate drug safety or purity. This test is for forensic use only.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Rapid Response™ Fentanyl (FYL) Test Strip is a rapid visual immunoassay for the qualitative detection of fentanyl at a cut-off of 200 ng/ml for liquid/powder or 20 ng/ml for urine.
- Parameter: FYL
- Calibrator: Fentanyl
- Cut-off: 200 ng/ml (Liquid/Powder), 20 ng/ml (Urine)
Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no control line).
Delivery information: Includes test strips, product insert.
Caution: This test does not evaluate drug safety or purity. This test is for forensic use only.
Product Family Options
Product Information
- DésignationTailleAvailabilityPrice
- Fentanyl (FYL) test strip100 testsKIT(100items)$225.00Kit of 100$180.62Supplier Number: FYL-1S48-100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: FYL-1S48-100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.