Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Pathfinder™ <i>Listeria</i> Broth

Pathfinder™ Listeria Broth

Listeria broth, 20×125 mm, 15 ml, PC tube, 20 per pack
Catalog # CA76414-754
Supplier #:  K321
Pathfinder™ <i>Listeria</i> Broth
Catalog # CA76414-754
Supplier:  
CAS Number:  
Pathfinder™ <i>Listeria</i> Broth
Pathfinder™ Listeria Broth

Listeria broth, 20×125 mm, 15 ml, PC tube, 20 per pack

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Pathfinder™ Listeria Broth is recommended for use with environmental samples from food processing environments or food contact surfaces as a selective and differential screening medium to detect and confirm the presence of Listeria spp. after cleaning.

  • Less steps, less time, less labor, less errors
  • Read your results as early as 30 h
  • After an additional incubation, Listeria spp. organisms will cause a fluorescent reaction
  • Leak-proof caps provide security during handling and transportation
  • Tubes have excellent impact and are shatter resistant

Eliminate false positives by using our confirmatory Listeria spp. environmental media!

Four Easy Steps!

1. Remove sampling device from container, such as EnviroMax® Environmental Sampling Swab (Cat. no. 89405-022), to swab the surface.
2. Aseptically pour all the contents of the Pathfinder™ Listeria Broth tube into the swab container.
3. Incubate at 37 °C for 30 h.
4. Read results.

Specifications

Specifications

  • Type de conditionnement:
    20 per pack
  • Désignation:
    Listeria broth
  • Taille:
    20×125 mm
  • Fill Volume:
    15 ml
  • Format:
    PC tube

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.