EMS Disc Punch for TEM Preparation, Electron Microscopy Sciences
Wide base specimen punch
  76468-304
 :  50125-10
EMS Disc Punch for TEM Preparation, Electron Microscopy Sciences
  76468-304
EMS Disc Punch for TEM Preparation, Electron Microscopy Sciences
EMS Disc Punch for TEM Preparation, Electron Microscopy Sciences
Wide base specimen punch
$6,713.80
Each
The EMS 310 Disc Punch is designed with 3 mm disc to prepare ductile metals and soft materials for TEM without mechanical distortion. Its solid construction and high tolerance design allows a maximum force to be used with difficult materials while maintaining specimen quality.
  • Preparation of TEM samples without distortion
  • Alignment fixture included for maximum yield and precise specimen location
  • Solid construction for long term use without loss of precision
A specimen alignment tool is included which guarantees maximum specimen yield and precise specimen location. After the disc is punched it is automatically selected from the die and collected in an easily removable tray.
Dimensions - 8 W×8 H×2.5 D"
Weight - 5 lbs.
  • Désignation:
    Wide base specimen punch