Small Pliers, Excelta
Supplier: Excelta
Jump to:
À propos de cet article
Three star quality small pliers made of carbon steel with flame hardened tips and feature dual leaf springs for smooth action.The grips are soft cushioned foam and are ESD safe.
4 Options Available Below
Product Details & Documents
Three star quality small pliers made of carbon steel with flame hardened tips and feature dual leaf springs for smooth action.The grips are soft cushioned foam and are ESD safe.
Product Family Options
Informations sur le produit
- TailleLongueurTip SizeTip StyleAvailabilityPrice
- Supplier Number: 42I
- Supplier Number: 43I
- Supplier Number: 44I
- Supplier Number: 47I