Copan Loop, Disposable, Rigid (hard) Plastic, Hardy Diagnostics
Product Details & Documents
Hexagonal loop shaft improves grip, while the ultra smooth loop surfaces allow for easy streaking.
- Loops are sterile
- Free of lubricants, oils, and electrostatic charges
: Packed in re-sealable plastic pouches.
Spécifications
- Color:Dark blue
- Désignation:Loop, disposable plastic
- Taille:10 µl
- Packaging:40 per pack/1000 per box
- Forme:Rigid (hard) plastic