Miltex® Scissors, Integra
Supplier:   Miltex
Miltex® Scissors, Integra
Supplier:  Miltex
Certificates
Miltex® Scissors, Integra
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Integra Miltex recognizes and meets a variety of clinical needs in the careful construction of these surgical scissors.
Whether dissecting, cutting tissue, suture or dressing material or performing any number of important functions, our finely crafted scissors perform as intended for the surgical task.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Blade Type
    Curvature
    Availability
    Price
  • Iris scissors, SuperCut
    116 mm (4⅝")
    Sharp/sharp
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $373.77
    $373.77
    Each
       $373.77
    Supplier Number: 5-SC-304
  • Iris scissors, SuperCut
    116 mm (4⅝")
    Sharp/sharp
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $405.67
    $405.67
    Each
       $405.67
    Supplier Number: 5-SC-306
  • Mayo scissors
    140 mm (5½")
    Blunt
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $385.37
    $385.37
    Each
       $385.37
    Supplier Number: 5-SC-120
  • Mayo scissors
    140 mm (5½")
    Blunt
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $402.41
    $402.41
    Each
       $402.41
    Supplier Number: 5-SC-122
  • Mayo scissors, SuperCut, one serrated blade
    172 mm (6¾")
    Blunt
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $421.70
    $421.70
    Each
       $421.70
    Supplier Number: 5-SC-126
  • Metzenbaum scissors, SuperCut
    179 mm (7")
    Blunt/Blunt
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $421.68
    $421.68
    Each
       $421.68
    Supplier Number: 5-SC-182
  • Metzenbaum scissors, SuperCut, delicate, one serrated blade
    115 mm (4½")
    Blunt/Blunt
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $397.20
    $397.20
    Each
       $397.20
    Supplier Number: 5-SC-284
  • Operating scissors, SuperCut
    145 mm (5¾")
    Sharp-blunt points
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $369.90
    $369.90
    Each
       $369.90
    Supplier Number: 5-SC-16
  • Operating scissors, SuperCut, one serrated blade
    145 mm (5¾")
    Sharp-blunt points
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $397.20
    $397.20
    Each
       $397.20
    Supplier Number: 5-SC-46