pMAL-c6T Vector | Avantor
pMAL-c6T Vector
pMAL-c6T vector, 10 μg (50 μl)
Catalog # CA76423-414
New Product
Fournisseur #:  N0378S
pMAL-c6T Vector
Catalog # CA76423-414
New Product
CAS Number:  
pMAL-c6T Vector
pMAL-c6T Vector
pMAL-c6T vector, 10 μg (50 μl)
CAS Number:  
Price
$251.00
Log in pour la disponibilité
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The pMAL-c6T Vector is designed to produce maltose-binding protein (MBP) fusions in the cytoplasm.
  • pMAL-c6T is the recommended vector for use with the NEBExpress MBP fusion and purification system
  • Fusion to MBP significantly enhances the solubility and proper folding of proteins in E. coli
  • Includes TEV Protease site, allowing protein to be cleaved from MBP using TEV Protease (NEB #P8112)
  • Two-step purification: amylose elution followed by TEV Protease cleavage and Ni resin isolation
The MBP has been engineered for tighter binding to amylose resin. The vector expresses an N-terminal his-tagged mal E gene followed by a multiple cloning site containing a TEV protease recognition sequence, allowing MBP to be cleaved from the protein of interest after purification.
Spécifications
Spécifications
  • Désignation:
    pMAL-c6T vector
  • Taille:
    10 μg (50 μl)