yDcpS | Avantor
yDcpS
4000 units
Catalog # CA76339-732
Fournisseur #:  M0463S
yDcpS
Catalog # CA76339-732
CAS Number:  
yDcpS
yDcpS
4000 units
CAS Number:  
Price
$466.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
yDcpS is the scavenging decapping enzyme from S. cerevisiae that hydrolyzes the phosphodiester bond between the gamma and beta phosphates of m7G-capped mRNA, producing diphosphorylated 5 ends and m7GMP. yDcpS is capable of decapping mRNAs of various lengths, decaps Cap0 and Cap1 with equal efficiency, and the diphosphorylated 5 end it produces is suitable for recapping with Vaccinia capping enzyme.
  • Provided at high enzyme concentration to facilitate complete decapping
  • Active on capped RNA of various sizes (tested on in vitro capped transcripts from 25 nt to 1400 nt)
  • Decapped RNA 5' ends can be recapped with Vaccinia capping system supports tagging of mRNA with desthiobiotin-labeled GTP
Spécifications
Spécifications
  • Taille:
    4000 units
  • Cat. No.:
    CA76339-732
  • Concentration:
    200000 units/ml