ContactUsLinkComponent
Luna Cell Ready Lysis Module, New England Biolabs
Luna Cell Ready Lysis Module, New England Biolabs
Catalog # CA76407-360
CAS Number:  
undefined
Luna Cell Ready Lysis Module, New England Biolabs
Catalog # CA76407-360
Supplier Number:  E3032S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Luna Cell ready lysis module
  • Size:
    100 reactions
  • Cat. no.:
    CA76407-360
Specifications
About this item
Cell cultures are often analyzed for gene expression or treatment responses as a proxy for a living organism. Traditionally, RNA is extracted and purified from treated cells via column-based or chemical methods. The Luna Cell Ready Lysis Module is a fast, convenient, and sensitive alternative for evaluating RNA expression levels without performing traditional RNA extraction steps.
  • Coordinated cell lysis, RNA release, and genomic DNA removal in a fast 15-minute protocol
  • Effective cell lysis preparation from 10 to 100000 cells across numerous cell lines
  • Minimal sample loss compared to alternative RNA extraction methods, especially at low cell input
  • Compatible with high-throughput applications: cell lysis can take place at room temperature
The module coordinates cell lysis, RNA release, and genomic DNA removal simultaneously, resulting in cell lysate ready for direct RT-qPCR analysis with Luna Universal One-Step RT-qPCR kits. Similar to other Luna products, the lysis buffer includes an inert blue tracking dye for visual assistance throughout the workflow.