Floor Marking Tape, Blank, Yellow, Accuform
Catalog # 76471-364
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Width:2"
- Color:Yellow
- Description:Floor marking tape, blank
- Length:108'
- Cat. no.:76471-364
Specifications
About this item
Marking tape is used to mark specific areas and promote safety by identifying potential hazard areas.
- Vinyl adheres to floors, walls, piping, posts, beams, handrails, stairways, machinery, and equipment
- Permanent self-adhesive back with no liner
Extremely durable floor marking tape is waterproof and wear-resistant, while offering minor resistance to acids, oils, grease, caustics, alkalis, and most solvents.