SP Bel-Art Stainless Steel Pharma Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These reusable, high-quality scoops can be used time and time again for powder and granule handling in pharmaceutical and food handling settings.
- Pharmaceutical 316 grade stainless steel
- Easy to clean seamless design to help prevent cross-contamination
- High salt and acid resistance, ideal for food sampling
- GMP compliant
- Autoclavable
Helps to meet sustainability goals when used in place of disposable scoops.
Frequently Bought Together