About this item
The StaticCare™ ESD Cut Resistant Gloves combine powerful cut resistance, comfort, and uniform ESD protection into one superior performing glove. The gloves are certified ANSI Cut Level 3 while maintaining softness, flexibility, and comfort.
- Multi-purpose electronics inspection and handling glove
- Typical ESD properties: <10⁷ ohms
- UHMWPE construction with conductive carbon
- Available uncoated, fingertip coated, or palm coated
- Finished knit wrist cuff
The StaticCare™ GL2500 series gloves provide superior ESD protection because conductive carbon threads are combined with the cut resistant fibers to provide uniform static decay rates.The gloves are constructed with 18 gauge Ultra High Molecular Weight Polyethylene (UHMWPE) thread which provides great cut protection, as well as outstanding abrasion and tear resistance. They are rated ANSI Cut Level 3, which offers a basic level protection against cuts or punctures.
A finished knitwrist cuff provides a snug fit to reduce slippage and keep debris out. Available with polyurethane coated finger tips or palms to improve grip while maintaining breathability. The gloves may be laundered for repeated use.
Specifications
- Catalog No:
- 76472-910
- 76472-932
- 76472-908
- 76472-930
- 76472-928
- 76472-906
- 76472-926
- 76472-904
- 76472-912
- 76472-916
- 76472-914
- 76472-934
- 76472-924
- 76472-902
- 76472-922
- 76472-900
- 76472-920
- 76472-918
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +