ContactUsLinkComponent
Nucleoside Digestion Mix
Nucleoside Digestion Mix
Catalog # CAMSPP-M0649S
CAS Number:  
undefined
Nucleoside Digestion Mix
Catalog # CAMSPP-M0649S
Supplier Number:  M0649S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    50 reactions
  • Cat. no.:
    CAMSPP-M0649S
Specifications
About this item
The Nucleoside Digestion Mix is a mixture of enzymes that provides a convenient one-step method to generate single nucleosides from DNA or RNA.
  • Convenient one-step protocol
  • Digests both DNA and RNA to single nucleosides
  • Low-glycerol formulation significantly reduces glycerol-induced ion suppression during MS analysis
Optimized for quantitative analysis by liquid chromatography-mass spectrometry (LC-MS), this reagent eliminates the need for sequential multi-step, time-consuming digestion protocols. The Nucleoside Digestion Mix digests ssDNA, dsDNA, DNA/RNA hybrids and RNA (except mRNA cap structures) containing epigenetically modified (m5C, hm5C, f5C, ca5C, m4C, m6A, etc.), unnatural, or damaged bases. Moreover, the low-glycerol formulation (<1%) significantly reduces glycerol-induced ion suppression during mass spectrometry analysis.