NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
The NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module uses a template switching method to generate full length cDNAs directly from single cells or 2 pg to 200 ng RNA.
- Experience unmatched detection of low abundance transcripts
- Rely on consistent transcript detection for a wide range of input amounts and sample types
- Observe uniform transcript coverage, regardless of input amount or sample type
- Use with a variety of RNA inputs, including cultured or primary cells, or total RNA
This unique workflow enables generation of high quality sequencing data from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance.
Frequently Bought Together