NEB 10-beta/Stable Outgrowth Medium, New England Biolabs
76196-414
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:NEB 10-beta/stable outgrowth medium
- Size:100 (4×25 ml)
- Cat. no.:76196-414
Outgrowth medium recommended for use with NEB 10-beta or NEB Stable Competent E. coli.
- Use to dilute the outgrowth prior to plating
Add NEB 10-beta/Stable Outgrowth Medium to freshly transformed NEB 10-beta or NEB Stable E. coli cells and incubate for up to 1 hour before plating on selective media.