VWR® Vertical Racks with Spring Lock
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
Jump to:
À propos de cet article
Vertical racks for mini 2" boxes with spring lock.
- Stainless steel
- Double welds
- Smoothed edges
- Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
7 Options Available Below
Product Details & Documents
Vertical racks for mini 2" boxes with spring lock.
- Stainless steel
- Double welds
- Smoothed edges
- Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Product Family Options
Informations sur le produit
- DésignationCapacitéProfondeurHauteurLargeurAvailabilityPrice
- Vertical rack with spring lock9× 2" mini boxes3¼"20¹⁄₁₆"3⅜"Log in pour la disponibilitéEach$108.73Supplier Number: MCF-9-2-S
- Vertical rack with spring lock10× 2" mini boxes3¼"22⁵⁄₁₆"3⅜"Log in pour la disponibilitéEach$113.79Supplier Number: MCF-10-2-S
- Supplier Number: MCF-11-2-S
- Supplier Number: MCF-12-2-S
- Supplier Number: MCF-13-2-S
- Vertical rack with spring lock14× 2" mini boxes3¼"31¹⁄₁₆"3⅜"Log in pour la disponibilitéEach$147.98Supplier Number: MCF-14-2-S
- Supplier Number: MCF-15-2-S