ContactUsLinkComponent
Citrus Clearing Solvent, Epredia
Catalog # CA72060-044
Supplier:  Thermo Scientific
CAS Number:  
Citrus Clearing Solvent, Epredia
Catalog # CA72060-044
Supplier:  Thermo Scientific
Supplier Number:  8301
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pack type:
    Poly Bottle
  • Description:
    Citrus Clearing Solvent
  • Size:
    1 gal.
  • Cat. no.:
    CA72060-044
  • Supplier No.:
    8301
Specifications
About this item
Replace xylene in both manual and automated system applications with Epredia™ Citrus Clearing Solvent.
  • D-limonene-based solvent that has a slight citrus scent
  • Higher flash point eliminates the need for storage in a flammable safety cabinet