Clear-Rite 3™ Clearant, Epredia
Clear-Rite 3™ Clearant, Epredia
Clear-Rite 3™ Clearant, Epredia
Clear-Rite 3™ Clearant, Epredia
Clear-Rite 3™ Clearant, Epredia
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series™ Clear-Rite 3™, a unique blend of isoparaffinic aliphatic hydrocarbons.
  • Very low odor; benzene-free
  • Use in all tissue processors and automatic stainers
  • Does not produce hard or brittle specimens, even after prolonged exposure
Product Family Options
  • Désignation
    Utilisation
    Type de conditionnement
    Taille
  • Clear-Rite 3®
    Lipid Extraction, Complete Deparaffinization and Clearing
    Poly Bottle
    1 gal.
    Case of 4 (0.25 .US Gal)
    Case of 4 (0.25 .US Gal) - $814.19
    $814.19
    Case of 4 (0.25 .US Gal)
       $814.19
     : 6901
  • Clear-Rite 3®
    Lipid Extraction, Complete Deparaffinization and Clearing
    Metal Pail
    5 gal.
    Each (5US Gal)
    Each (5US Gal) - $1,013.13
    $1,013.13
    Each (5US Gal)
       $1,013.13
     : 6905