AffinityScript cDNA Synthesis Kits, Agilent Technologies | Avantor
AffinityScript cDNA Synthesis Kits, Agilent Technologies
AffinityScript cDNA Synthesis Kits, Agilent Technologies
Certificates
AffinityScript cDNA Synthesis Kits, Agilent Technologies
AffinityScript cDNA Synthesis Kits, Agilent Technologies
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Provides the highest efficiency of conversion of RNA to cDNA, fully optimized for qPCR applications.
  • Optimized for real-time quantitative PCR (qPCR) application;Produces cDNA in 15 min with linear detection from 3 pg to 3 µg total RNA
  • Creates up to 12 kb cDNA in 25 min or less
  • Available as a first-strand synthesis kit and a two-step QRT-PCR master mix format
The AffinityScript qPCR cDNA Synthesis Kit offers efficient conversion of RNA to cDNA. Fully optimized for real-time quantitative PCR (qPCR) applications, the kit utilizes our qPCR-grade AffinityScript Multiple Temperature Reverse Transcriptase. It includes a buffer system optimized for real-time qPCR.
The streamlined protocol produces cDNA in 15 min with linear detection from 3 pg to 3 µg of total RNA. The easy-to-use process generates up to 12 kb cDNA in 25 min or less. Available as a first-strand synthesis kit and a two-step QRT-PCR master mix format, using SYBR Green or probe-based detection.
The AffinityScript Multiple Temperature cDNA synthesis kit contains Agilent’s popular, highly thermostable, qPCR-grade AffinityScript Multiple Temperature Reverse Transcriptase for robust yields, from 37 to 60 °C, with nearly full activity between 45 to 55 °C. With the AffinityScript multiple temperature cDNA synthesis kit, Agilent offers enzymes with a broad thermal profile.
The Agilent AffinityScript Multiple Temperature Reverse Transcriptase (RT) system is optimized to produce high cDNA yields over the broadest temperature range. During analysis the multi-temp RT device also ensures high affinity to primer-template complexes. cDNA can be synthesized in amounts up to 20 kb.
The system’s multi-temperature capability allows you to change RT reaction temperature without having to change the reverse transcriptase, for a faster and more efficient analysis. In addition, when amplifying low RNA input amounts and resolving RNA secondary structures it delivers high cDNA yields, and can operate effectively at temperatures up to 55 °C.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Details
    No. of Reactions
    Availability
    Price
  • AffinityScript qPCR cDNA synthesis Kit
    Kit delivers fast, efficient cDNA synthesis for qRT-PCR. Produces cDNA in 15 minutes with linear detection from 3 pg to 3 µg total RNA. Creates up to 12 kb cDNA in 25 min or less
    50
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $470.36
    $470.36
    Each
       $470.36
    La quantité minimale de commande est de 1
    Supplier Number: 600559
  • AffinityScript cDNA Synthesis Kit
    Kit utilizes our qPCR-grade AffinityScript Multiple Temperature Reverse Transcriptase. It includes a buffer system optimized for real-time qPCR
    50
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $402.72
    $402.72
    Each
       $402.72
    Supplier Number: 200436
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • AffinityScript multiple temperature reverse transcriptase
    Engineered to be highly thermostable, allowing you to reverse transcribe at your preferred reaction temperature. Achieve a robust cDNA yield with even GC-rich sequences. Increased activity at all temperatures in a broad operating range, from 37 to 55 °C. The system is able to synthesize cDNA up to 20 kb
    50
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $281.59
    $281.59
    Each
       $281.59
    La quantité minimale de commande est de 1
    Supplier Number: 600107
  • AffinityScript multiple temperature reverse transcriptase
    Engineered to be highly thermostable, allowing you to reverse transcribe at your preferred reaction temperature. Achieve a robust cDNA yield with even GC-rich sequences. Increased activity at all temperatures in a broad operating range, from 37 to 55 °C. The system is able to synthesize cDNA up to 20 kb
    200
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,002.26
    $1,002.26
    Each
       $1,002.26
    La quantité minimale de commande est de 1
    Supplier Number: 600109
  • AffinityScript one-step RT-PCR Kit
    Reduced PCR cycling times and a one-tube, master mix format. Combined performance of Herculase II fusion DNA polymerase and the ArchaeMaxx PCR enhancing factor, delivering superior PCR yield and reliability. Robust performance for difficult and GC-rich targets. Optimized for target lengths up to 9.5 kb.
    100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - $661.11
    $661.11
    Kit of 2
       $661.11
    La quantité minimale de commande est de 1
    Supplier Number: 600188
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.