AccuScript High-Fidelity Reverse Transcriptase, Agilent Technologies
À propos de cet article
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
- MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
- High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0.1 to 10 ng of poly(A)+ RNA
- Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3.7 times more accuracy than other commercially available RTs and 6.6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
: For research use only. Not for use in diagnostic procedures.
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
- MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
- High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0.1 to 10 ng of poly(A)+ RNA
- Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3.7 times more accuracy than other commercially available RTs and 6.6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
: For research use only. Not for use in diagnostic procedures.
Product Family Options
Informations sur le produit
- DésignationDetailsNo. of ReactionsAvailabilityPrice
- AccuScript high-fidelity 1st strand cDNA synthesis kitThe kit delivers the highest accuracy and offers you greater flexibility to use the cDNA in your choice of downstream applications such as PCR amplification or real-time PCR quantification50Log in pour la disponibilitéEach$427.94Supplier Number: 200820L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- AccuScript reverse transcriptaseEngineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance50Log in pour la disponibilitéEach$315.93Supplier Number: 600089L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- AccuScript reverse transcriptaseEngineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance200Discontinued-Supplier Number: -
- AccuScript high-Fidelity RT-PCR kitHigh fidelity reverse transcriptase for long target lengths. Ideal for low RNA input and high specificity. Shorter PCR extension times with PfuUltra II with high accuracy. Kit includes dNTP, dTT, oligodT and random primers. Capable of amplifying cDNA of 0.1 - 9.6 kb in length ;10 – 1000 ng of total RNA or 0.1 – 10 ng of poly(A)+ RNA input50Discontinued-Supplier Number: -
- AccuScript PfuUltra II RT-PCR kitThe AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput50Discontinued-Supplier Number: -L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.