Electrode Filling Solution, 3 M KCl Electrolyte
Potassium chloride solution 3 M KCl, 250 ml, Bottle, Barium, fluoride, calcium, sodium, lithium
Supplier: Mettler Toledo
Fournisseur #: 51350072
Electrode Filling Solution, 3 M KCl Electrolyte
Catalog # 76359-830
Supplier: Mettler Toledo
CAS Number:
Jump to:
Product Details & Documents
Mettler-Toledo offers electrolytes for every application. Fill-up your electrode with correct and fresh electrolyte for a precise measurement.
Sensor used: Reference cell, combined electrode
Parameter: pH and ISE
Documents
Certificates
Recherche par numéro de lotSpécifications
Spécifications
- Type de conditionnement:Bottle
- Désignation:Potassium chloride solution 3 M KCl
- Taille:250 ml
- Ion:Barium, fluoride, calcium, sodium, lithium
- Cat. No.:76359-830