SP Bel-Art Sterileware® Powder Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These sterile, wide stem funnels are ideal for transferring powders or granules using a sterile sampling tool into a sterile container.
- 60° angle and large bore stem help minimize bridging of powder
- FDA grade polypropylene; safe for food, drug and cosmetic use
- Gamma irradiated sterile (SAL 10⁻⁶)
- Individually bagged; ready for one time use and disposal
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Frequently Bought Together