CIP 200® Acid-Based Process and Research Cleaners
Supplier:   Steris
CIP 200® Acid-Based Process and Research Cleaners
Supplier:  Steris
Certificates
CIP 200® Acid-Based Process and Research Cleaners
CIP 200® Acid-Based Process and Research Cleaners
6 Options Available Below
Product Details & Documents
CIP 200® Acid Detergent is a proprietary blend of phosphoric acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming detergent is ideal for use in CIP, COP and manual applications for a variety of applications including derouging and passivation.
  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Global Availability - and manufactured with biodegradable surfactants to support harmonization of cleaning processes
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
CIP 200® Acid-Based Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
Product Family Options
Informations sur le produit
  • Taille
    Packaging
    Availability
    Price
  • 3.8 L (1 gal.)
    Plastic bottle, 4/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $992.00
    $992.00
    Case of 4
       $992.00
    Supplier Number: 1D2308
  • 19 L (5 gal.)
    Plastic pail
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $896.00
    $896.00
    Each(1/Itm/Bx)
       $896.00
    Supplier Number: 1D2305
  • 114 L (30 gal.)
    Plastic drum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,230.00
    $3,230.00
    Each
       $3,230.00
    Supplier Number: 1D2302
  • 208 L (55 gal.)
    Plastic drum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,100.00
    $9,100.00
    Each
       $9,100.00
    Supplier Number: 1D2301
  • 757 L (200 gal.)
    Plastic tote
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17,160.00
    $17,160.00
    Each
       $17,160.00
    Supplier Number: 1D23D9
  • 1136 L (300 gal.)
    Plastic tote
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22,611.60
    $22,611.60
    Each
       $22,611.60
    Supplier Number: 1D23D3