VWR® Sliding Tray Racks
Fournisseur: VWR International
Certificats
À propos de cet article
Sliding tray rack for 2" boxes.
- Stainless steel
- Double welds
- Smoothed edges
- Lifetime warranty
These freezer racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Frequently Bought Together