VWR® Freezer Drawer Racks
VWR® Freezer Drawer Racks
VWR® Freezer Drawer Racks
VWR® Freezer Drawer Racks
Drawer rack for 2" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
These freezer racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Product Family Options
  • Désignation
    Capacité
    Profondeur
    Hauteur
    Largeur
  • Freezer drawer rack
    8× 2" boxes
    11⁷⁄₁₆"
    9⁷⁄₁₆"
    5½"
    Each
    Each - $195.46
    $195.46
    Each
       $195.46
  • Freezer drawer rack
    10× 2" boxes
    11⁷⁄₁₆"
    11¹³⁄₁₆"
    5½"
    Each
    Each - $265.51
    $265.51
    Each
       $265.51
  • Freezer drawer rack
    12× 2" boxes
    11⅜"
    14⅛"
    5½"
    Each
    Each - $278.37
    $278.37
    Each
       $278.37
  • Freezer drawer rack
    12× 2" boxes
    22⁵⁄₁₆"
    5½"
    7⅛"
    Each
    Each - $323.34
    $323.34
    Each
       $323.34
  • Freezer drawer rack
    15× 2" boxes
    16¾"
    11¹³⁄₁₆"
    5½"
    Each
    Each - $445.55
    $445.55
    Each
       $445.55
  • Freezer drawer rack
    15× 2" boxes
    27"
    7⅛"
    5½"
    Each
    Each - $337.73
    $337.73
    Each
       $337.73
  • Freezer drawer rack
    18× 2" boxes
    16¾"
    14⅛"
    5½"
    Each
    Each - $401.61
    $401.61
    Each
       $401.61
  • Freezer drawer rack
    28× 2" boxes
    22⁵⁄₁₆"
    16½"
    5½"
    Each
    Each - $685.36
    $685.36
    Each
       $685.36
  • Freezer drawer rack
    30× 2" boxes
    27"
    14⅝"
    5½"
    Each
    Each - $696.04
    $696.04
    Each
       $696.04