VWR® Vertical Racks
Supplier:   VWR International
VWR® Vertical Racks
Supplier:  VWR International
Certificates
VWR® Vertical Racks
VWR® Vertical Racks
5 Options Available Below
Product Details & Documents
Vertical freezer racks for 2" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Product Family Options
Product Information
  • Désignation
    Capacité
    Profondeur
    Hauteur
    Largeur
    Availability
    Price
  • Vertical rack
    3× 2" mini boxes
    3¼"
    6⅞"
    3⅜"
    Log in for availability
    Log in for availability
    Each
    Each - $37.46
    $37.46
    New Customer Promotional Price
    Each
       $37.46
  • Vertical rack
    12× 2" mini boxes
    3¼"
    26⅝"
    3⅜"
    Log in for availability
    Log in for availability
    Each
    Each - $104.71
    $104.71
    New Customer Promotional Price
    Each
       $104.71
  • Vertical rack
    13× 2" mini boxes
    3¼"
    28¹⁄₃"
    3⅜"
    Log in for availability
    Log in for availability
    Each
    Each - $118.31
    $118.31
    New Customer Promotional Price
    Each
       $118.31
  • Vertical rack
    14× 2" mini boxes
    3¼"
    31¹⁄₁₆"
    3⅜"
    Log in for availability
    Log in for availability
    Each
    Each - $130.59
    $130.59
    New Customer Promotional Price
    Each
       $130.59
  • Vertical rack
    15× 2" mini boxes
    3¼"
    33¼"
    3⅜"
    Log in for availability
    Log in for availability
    Each
    Each - $155.13
    $155.13
    New Customer Promotional Price
    Each
       $155.13